Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VIPAS39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193314.1 | NP_001180243.1 | 12 | 1352 | ||
NM_001193315.1 | NP_001180244.1 | 12 | 952 | |||
NM_001193316.1 | NP_001180245.1 | 11 | 805 | |||
NM_001193317.1 | NP_001180246.1 | 12 | 1129 | |||
NM_022067.3 | NP_071350.2 | 13 | 1223 | |||
XM_011537066.1 | XP_011535368.1 | 11 | 850 | |||
XM_017021579.1 | XP_016877068.1 | 11 | 984 | |||
XM_017021580.1 | XP_016877069.1 | 12 | 943 | |||
XM_017021581.1 | XP_016877070.1 | 12 | 1077 | |||
GenBank mRNA | AK022769.1 | - | 12 | 900 | ||
AK022925.1 | - | 12 | 878 | |||
AK024661.1 | - | 1 | 48 | |||
AK098254.1 | - | 12 | 1313 | |||
AK222706.1 | - | 12 | 898 | |||
AK298354.1 | - | 11 | 751 | |||
BC015054.1 | - | 13 | 1144 | 3920459 | ||
CR457294.1 | - | 11 | 806 |