Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VIPAS39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193314.1 | NP_001180243.1 | 17 | 1742 | ||
NM_001193315.1 | NP_001180244.1 | 17 | 1342 | |||
NM_001193316.1 | NP_001180245.1 | 16 | 1195 | |||
NM_001193317.1 | NP_001180246.1 | 17 | 1519 | |||
NM_022067.3 | NP_071350.2 | 18 | 1613 | |||
XM_011537066.1 | XP_011535368.1 | 16 | 1240 | |||
XM_017021579.1 | XP_016877068.1 | 16 | 1374 | |||
GenBank mRNA | AK022769.1 | - | 17 | 1290 | ||
AK022925.1 | - | 17 | 1268 | |||
AK024661.1 | - | 6 | 438 | |||
AK098254.1 | - | 17 | 1703 | |||
AK222706.1 | - | 17 | 1288 | |||
AK297388.1 | - | 17 | 1444 | |||
AK298354.1 | - | 16 | 1141 | |||
BC015054.1 | - | 18 | 1534 | 3920459 | ||
CR457294.1 | - | 16 | 1196 |