Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: GALNT11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304514.1 | NP_001291443.1 | 2, 3 | 335 | ||
NM_022087.3 | NP_071370.2 | 3, 4 | 668 | |||
XM_006716082.2 | XP_006716145.1 | 3, 4 | 579 | |||
XM_006716083.2 | XP_006716146.1 | 3, 4 | 547 | |||
XM_006716084.2 | XP_006716147.1 | 3, 4 | 511 | |||
XM_017012499.1 | XP_016867988.1 | 3, 4 | 649 | |||
GenBank mRNA | AK025287.1 | - | 3, 4 | 502 | ||
AK124934.1 | - | 5, 6 | 856 | |||
AK298911.1 | - | 2, 3 | 258 | |||
BC059377.1 | - | 4, 5 | 774 | |||
Y12434.3 | - | 2, 3 | 419 |