Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF335 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022095.3 | NP_071378.1 | 23 | 3701 | ||
XM_005260504.4 | XP_005260561.1 | 23 | 4414 | |||
XM_017028012.1 | XP_016883501.1 | 22 | 3472 | |||
GenBank mRNA | AF395833.1 | - | 23 | 3678 | ||
AK026157.1 | - | 15 | 2216 | |||
AK074214.1 | - | 8 | 1336 | |||
AK122963.1 | - | 6 | 1892 | |||
AK296989.1 | - | 20 | 3272 | |||
BC041331.2 | - | 20 | 2916 | |||
BC142981.1 | - | 20 | 2916 |