Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKEF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303472.1 | NP_001290401.1 | 7 | 1787 | ||
NM_022096.5 | NP_071379.3 | 7 | 1805 | |||
NM_198798.2 | NP_942093.1 | 6 | 1741 | |||
XM_005260792.2 | XP_005260849.1 | 5 | 1274 | |||
GenBank mRNA | AK025322.1 | - | 1 | 378 | ||
AK097689.1 | - | 6 | 1674 | |||
BC022878.2 | - | 7 | 1715 | 4821619 |