Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: XPNPEP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022098.3 | NP_071381.1 | 9 | 1443 | ||
GenBank mRNA | AK313770.1 | - | 9 | 1404 | ||
AL365514.1 | - | 9 | 1426 | |||
AL834310.1 | - | 10 | 1485 | |||
BC001681.2 | - | 9 | 1410 | 2823656 | ||
BC004989.1 | - | 9 | 1410 | 2823656 | ||
BX648018.1 | - | 9 | 1438 | |||
CR456442.1 | - | 9 | 1379 | |||
CR457396.1 | - | 9 | 1342 |