Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DMRTB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033067.2 | NP_149056.1 | 2 | 647 | ||
GenBank mRNA | AJ291671.1 | - | 2 | 574 | ||
AK057273.1 | - | 2 | 647 | |||
AK302085.1 | - | 2 | 236 | |||
BC029566.1 | - | 2 | 163 | 4836894 | ||
KU178739.1 | - | 2 | 133 |