Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CPB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278541.1 | NP_001265470.1 | 7 | 702 | ||
NM_001872.4 | NP_001863.3 | 8 | 813 | |||
XM_017020393.1 | XP_016875882.1 | 8 | 756 | |||
GenBank mRNA | AB011969.1 | - | 7 | 654 | ||
AK290829.1 | - | 8 | 761 | |||
BC007057.1 | - | 8 | 770 | 3934520 | ||
BT006936.1 | - | 8 | 746 | |||
M75106.1 | - | 8 | 765 |