Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PRDM15 SIRNA Assays ›
Gene Symbol:
PRDM15
Gene Name:
PR domain 15
Gene Aliases:
C21orf83, PFM15, ZNF298
Chromosome Location:
Chr.21: 41798225 - 41879482 on Build GRCh38
UniGene ID:
Hs.473893
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s34351
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PRDM15
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001040424.2
NP_001035514.1
21
2740
NM_001282934.1
NP_001269863.1
22
2800
NM_022115.4
NP_071398.3
28
3727
NR_104257.1
22
2748
NR_104258.1
21
2628
NR_104260.1
21
2595
NR_135464.1
21
2736
XM_006724039.2
XP_006724102.1
21
2582
XM_006724040.2
XP_006724103.1
20
2522
XM_011529674.1
XP_011527976.1
21
2612
XM_011529675.1
XP_011527977.1
22
2656
XM_011529676.2
XP_011527978.1
22
5065
XM_011529677.2
XP_011527979.1
22
2741
XM_011529678.2
XP_011527980.1
22
2785
XM_011529679.2
XP_011527981.1
22
3118
XM_011529681.2
XP_011527983.1
22
4022
XM_011529683.1
XP_011527985.1
27
3443
XM_017028425.1
XP_016883914.1
21
2595
XM_017028426.1
XP_016883915.1
21
5004
GenBank mRNA
AB051812.1
-
21
2731
AB051813.1
-
22
2791
AB051814.1
-
22
2739
AB051815.1
-
21
2619
AB209162.1
-
1
4675
AF426259.1
-
1
188
AF426260.1
-
1
188
AY063456.1
-
3
211
AY078498.2
-
28
3727
BC067102.1
-
21
2595
5925140
HY146343.1
-
2
123
Back To Top
More Information
Panther Classification:
Molecular Function -
C2H2 zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
methylation
RNA polymerase II regulatory region sequence-specific DNA binding
transcription factor activity, sequence-specific DNA binding
methyltransferase activity
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-648b49bbc4-xm8cg:80/10.239.167.213:80.
git-commit: cd89e1917689fc5870ae005717bc737506e58003
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.17.2-2024.10.99-1.0