Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: MMP27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022122.2 | NP_071405.2 | 10 | 1394 | ||
XM_011542948.2 | XP_011541250.1 | 9 | 1313 | |||
XM_011542949.2 | XP_011541251.1 | 9 | 1280 | |||
XM_011542950.2 | XP_011541252.1 | 9 | 1140 | |||
XM_017018120.1 | XP_016873609.1 | 10 | 1439 | |||
GenBank mRNA | AF195192.1 | - | 10 | 1377 | ||
AY358752.1 | - | 10 | 1394 | |||
BX102293.1 | - | 6 | 642 |