Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NPAS3 SIRNA Assays ›
Gene Symbol:
NPAS3
Gene Name:
neuronal PAS domain protein 3
Gene Aliases:
MOP6, PASD6, bHLHe12
Chromosome Location:
Chr.14: 32934785 - 33804176 on Build GRCh38
UniGene ID:
Hs.657892
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s34382
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NPAS3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001164749.1
NP_001158221.1
8, 9
1118
NM_001165893.1
NP_001159365.1
7, 8
1028
NM_022123.2
NP_071406.1
7, 8
1022
NM_173159.2
NP_775182.1
8, 9
1079
XM_005267991.3
XP_005268048.2
8, 9
1223
XM_005267992.3
XP_005268049.2
8, 9
1205
XM_011537067.2
XP_011535369.1
9, 10
1219
XM_011537069.2
XP_011535371.2
8, 9
1223
XM_011537070.2
XP_011535372.1
8, 9
1123
XM_011537071.2
XP_011535373.2
7, 8
1118
XM_011537072.2
XP_011535374.1
7, 8
1077
XM_017021582.1
XP_016877071.1
9, 10
1274
XM_017021583.1
XP_016877072.1
9, 10
1274
XM_017021584.1
XP_016877073.1
8, 9
1184
XM_017021585.1
XP_016877074.1
7, 8
1133
XM_017021586.1
XP_016877075.1
7, 8
4030
XM_017021587.1
XP_016877076.1
7, 8
3272
XM_017021588.1
XP_016877077.1
8, 9
1220
GenBank mRNA
AB054575.1
-
7, 8
1009
AB054576.1
-
8, 9
1015
AB055962.1
-
8, 9
1054
AF164438.1
-
7, 8
1033
AY157302.1
-
7, 8
1022
KJ534893.1
-
7, 8
958
KJ534894.1
-
9, 10
1054
KJ534895.1
-
7, 8
964
KJ534896.1
-
7, 8
958
KJ534897.1
-
8, 9
1009
KJ534898.1
-
7, 8
964
KJ535019.1
-
8, 9
1015
Back To Top
More Information
Panther Classification:
Molecular Function -
basic helix-loop-helix transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
developmental process
positive regulation of transcription, DNA-templated
RNA polymerase II transcription factor activity, sequence-specific DNA binding
DNA binding
protein dimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-46vz9:80/10.239.163.144:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0