Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC28A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199633.1 | NP_001186562.1 | 14 | 1767 | ||
NM_022127.2 | NP_071410.1 | 15 | 1713 | |||
NR_037638.2 | 15 | 1943 | ||||
XM_011518905.2 | XP_011517207.1 | 14 | 1837 | |||
XM_011518906.2 | XP_011517208.1 | 15 | 1933 | |||
XM_011518907.2 | XP_011517209.1 | 13 | 1652 | |||
XM_011518908.2 | XP_011517210.1 | 9 | 1672 | |||
GenBank mRNA | AF305210.1 | - | 15 | 1713 | ||
AK292849.1 | - | 14 | 1766 | |||
AK304406.1 | - | 15 | 1923 | |||
AK314039.1 | - | 14 | 1747 | |||
BC093821.1 | - | 14 | 1650 | 7939666 | ||
BC093823.1 | - | 14 | 1650 | 7939668 |