Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPB41L4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022140.3 | NP_071423.3 | 17 | 1711 | ||
XM_011543530.1 | XP_011541832.1 | 17 | 1888 | |||
XM_011543531.2 | XP_011541833.1 | 17 | 1888 | |||
XM_011543532.1 | XP_011541834.1 | 17 | 1888 | |||
XM_011543533.1 | XP_011541835.1 | 16 | 1804 | |||
XM_017009689.1 | XP_016865178.1 | 16 | 1804 | |||
GenBank mRNA | AB030240.1 | - | 17 | 1443 | ||
AK096057.1 | - | 6 | 393 | |||
BC114632.1 | - | 17 | 1537 | |||
BC114942.1 | - | 17 | 1532 | |||
CB048933.1 | - | 4 | 266 |