Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CENPK SIRNA Assays ›
Gene Symbol:
CENPK
Gene Name:
centromere protein K
Gene Aliases:
AF5alpha, CENP-K, FKSG14, P33, Solt
Chromosome Location:
Chr.5: 65488919 - 65563194 on Build GRCh38
UniGene ID:
Hs.529778
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s34446
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CENPK
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001267038.1
NP_001253967.1
9
728
NM_022145.4
NP_071428.2
9
762
XM_011543534.2
XP_011541836.1
9
706
XM_011543535.2
XP_011541837.1
9
824
XM_011543536.2
XP_011541838.1
10
984
XM_011543537.2
XP_011541839.1
9
740
XM_011543538.2
XP_011541840.1
8
607
XM_011543539.2
XP_011541841.1
8
689
XM_011543540.2
XP_011541842.1
9
706
XM_011543541.2
XP_011541843.1
6
508
XM_011543542.2
XP_011541844.1
6
628
XM_017009690.1
XP_016865179.1
9
967
XM_017009691.1
XP_016865180.1
9
718
XM_017009692.1
XP_016865181.1
9
936
XM_017009693.1
XP_016865182.1
9
706
XM_017009694.1
XP_016865183.1
10
935
XM_017009695.1
XP_016865184.1
8
604
XM_017009696.1
XP_016865185.1
9
706
XM_017009697.1
XP_016865186.1
8
638
XM_017009698.1
XP_016865187.1
7
558
XM_017009699.1
XP_016865188.1
9
740
XM_017009700.1
XP_016865189.1
8
3072
XM_017009701.1
XP_016865190.1
7
703
GenBank mRNA
AF315941.1
-
9
718
AY009151.1
-
7
592
BC005400.1
-
9
718
3839409
BC008504.1
-
9
762
3511065
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
sister chromatid cohesion
CENP-A containing nucleosome assembly
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0