Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C10orf54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022153.1 | NP_071436.1 | 5, 6 | 854 | ||
GenBank mRNA | AF289594.1 | - | 1290 | |||
AK024449.1 | - | 5, 6 | 772 | |||
AY265805.1 | - | 5, 6 | 769 | |||
AY358379.1 | - | 5, 6 | 854 | |||
BC020568.1 | - | 5, 6 | 726 | 4158410 | ||
BC111048.1 | - | 5, 6 | 702 | |||
BC127257.1 | - | 2, 3 | 132 | |||
EF506490.1 | - | 5, 6 | 766 |