Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADGRL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022159.3 | NP_071442.2 | 12, 13 | 1900 | ||
GenBank mRNA | AB032017.1 | - | 12, 13 | 1861 | ||
AF192403.1 | - | 10, 11 | 1561 | |||
AK129766.1 | - | 3, 4 | 352 | |||
AK172750.1 | - | 9, 10 | 1379 | |||
AK292894.1 | - | 12, 13 | 1793 | |||
AK313738.1 | - | 12, 13 | 1899 | |||
AY358360.1 | - | 12, 13 | 1767 | |||
BC025721.1 | - | 12, 13 | 1766 | 5229055 |
Set Membership: |
![]() |