Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPEP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022355.3 | NP_071750.1 | 2 | 282 | ||
NR_136706.1 | 3 | 759 | ||||
XM_005256090.4 | XP_005256147.2 | 1 | 1239 | |||
XM_011523266.1 | XP_011521568.1 | 2 | 225 | |||
XM_011523268.2 | XP_011521570.1 | 1 | 1239 | |||
XM_011523271.2 | XP_011521573.1 | 1 | 1239 | |||
XM_011523272.2 | XP_011521574.1 | 2 | 323 | |||
XM_017023541.1 | XP_016879030.1 | 2 | 259 | |||
XM_017023542.1 | XP_016879031.1 | 1 | 1239 | |||
XM_017023546.1 | XP_016879035.1 | 2 | 259 | |||
GenBank mRNA | AJ295149.1 | - | 2 | 217 | ||
AK092884.1 | - | 3 | 759 | |||
AK298003.1 | - | 2 | 272 | |||
AK315090.1 | - | 1 | 259 | |||
AY358454.1 | - | 1 | 43 | |||
BC024021.1 | - | 2 | 266 | 4616323 | ||
BP299049.1 | - | 2 | 262 | |||
DA677374.1 | - | 2 | 225 | |||
DB038910.1 | - | 1 | 275 |