Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZSCAN31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135215.1 | NP_001128687.1 | 8 | 1548 | ||
NM_001135216.1 | NP_001128688.1 | 4 | 1044 | |||
NM_001243241.1 | NP_001230170.1 | 4 | 969 | |||
NM_001243242.1 | NP_001230171.1 | 4 | 666 | |||
NM_001243243.1 | NP_001230172.1 | 3 | 568 | |||
NM_001243244.1 | NP_001230173.1 | 3 | 557 | |||
NM_030899.4 | NP_112161.3 | 4 | 1033 | |||
NM_145909.2 | NP_665916.1 | 8 | 1170 | |||
XM_005249295.1 | XP_005249352.1 | 4 | 895 | |||
XM_005249296.4 | XP_005249353.1 | 5 | 5170 | |||
XM_011514807.2 | XP_011513109.1 | 6 | 5220 | |||
XM_011514808.2 | XP_011513110.1 | 4 | 5016 | |||
XM_011514809.1 | XP_011513111.1 | 4 | 880 | |||
XM_011514811.2 | XP_011513113.1 | 4 | 1395 | |||
XM_011514812.2 | XP_011513114.1 | 4 | 921 | |||
XM_011514813.2 | XP_011513115.1 | 4 | 1384 | |||
XM_017011196.1 | XP_016866685.1 | 6 | 5308 | |||
GenBank mRNA | AF513019.1 | - | 8 | 1170 | ||
AK027060.1 | - | 1 | 59 | |||
AK057702.1 | - | 8 | 1170 | |||
AK095261.1 | - | 8 | 1548 | |||
AK225382.1 | - | 1 | 62 | |||
AL833416.1 | - | 3 | 328 | |||
AL833617.1 | - | 4 | 1045 | |||
BC008490.1 | - | 4 | 807 | 4248596 | ||
BC065241.1 | - | 3 | 338 | 6016003 | ||
DA875036.1 | - | 3 | 557 | |||
DA879276.1 | - | 4 | 425 |