Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XPO4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022459.4 | NP_071904.4 | 23 | 3320 | ||
XM_005266502.1 | XP_005266559.1 | 24 | 3475 | |||
XM_005266503.3 | XP_005266560.1 | 20 | 2978 | |||
XM_005266504.3 | XP_005266561.1 | 17 | 2847 | |||
XM_011535197.1 | XP_011533499.1 | 24 | 3484 | |||
XM_011535198.1 | XP_011533500.1 | 22 | 3162 | |||
XM_011535199.1 | XP_011533501.1 | 21 | 3125 | |||
XM_011535200.2 | XP_011533502.1 | 21 | 4575 | |||
XM_017020704.1 | XP_016876193.1 | 19 | 3116 | |||
XM_017020705.1 | XP_016876194.1 | 20 | 4352 | |||
XM_017020706.1 | XP_016876195.1 | 17 | 2471 | |||
GenBank mRNA | AB051508.1 | - | 23 | 3283 | ||
AK023108.1 | - | 15 | 2276 | |||
AK057924.1 | - | 23 | 3290 | |||
AL831819.2 | - | 6 | 623 | |||
BC042504.1 | - | 20 | 2856 | |||
BC113571.1 | - | 22 | 3223 | 8322623 | ||
BC113573.1 | - | 22 | 3223 | 8322625 | ||
BX647843.1 | - | 14 | 2092 |