Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDST4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022569.2 | NP_072091.1 | 2 | 962 | ||
GenBank mRNA | AB036429.1 | - | 2 | 962 | ||
AK289670.1 | - | 2 | 940 |