Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENGASE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042573.2 | NP_001036038.1 | 5 | 711 | ||
XM_006722017.3 | XP_006722080.1 | 2 | 232 | |||
XM_006722019.3 | XP_006722082.1 | 5 | 836 | |||
XM_017024961.1 | XP_016880450.1 | 4 | 553 | |||
GenBank mRNA | AF512564.1 | - | 5 | 703 | ||
AK025518.1 | - | 5 | 711 | |||
AK301115.1 | - | 4 | 560 | |||
AL110283.1 | - | 1 | 34 |