Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXL17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163315.2 | NP_001156787.2 | 7 | 2212 | ||
XM_005272048.4 | XP_005272105.1 | 7 | 2206 | |||
XM_011543574.2 | XP_011541876.1 | 7 | 2206 | |||
XM_011543575.2 | XP_011541877.1 | 7 | 2206 | |||
GenBank mRNA | AB533280.1 | - | 8 | 1132 | ||
AK126722.1 | - | 8 | 1400 | |||
AL133602.1 | - | 6 | 813 | |||
BC126144.1 | - | 7 | 938 | 8991860 | ||
BC126146.1 | - | 7 | 938 | 8991862 |