Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATA20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258372.1 | NP_001245301.1 | 2 | 320 | ||
NM_001258373.1 | NP_001245302.1 | 3 | 423 | |||
NM_022827.3 | NP_073738.2 | 3 | 368 | |||
GenBank mRNA | AK025000.1 | - | 2 | 217 | ||
AK092731.1 | - | 3 | 353 | |||
AK125807.1 | - | 5 | 1098 | |||
AK225189.1 | - | 2 | 217 | |||
AK291805.1 | - | 3 | 311 | |||
AK302850.1 | - | 2 | 267 | |||
AK308616.1 | - | 3 | 493 | |||
AK310416.1 | - | 2 | 212 | |||
BC025255.1 | - | 3 | 256 | 5019010 | ||
BC065526.1 | - | 3 | 310 | 6149633 | ||
BC111029.1 | - | 2 | 225 | 5245538 | ||
BM787432.1 | - | 2 | 283 | |||
CR936769.1 | - | 2 | 238 | |||
DA295930.1 | - | 3 | 383 | |||
DA489234.1 | - | 3 | 368 | |||
DQ238597.1 | - | 2 | 286 | |||
GQ472220.1 | - | 2 | 225 |