Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATA20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258372.1 | NP_001245301.1 | 4 | 535 | ||
NM_001258373.1 | NP_001245302.1 | 5 | 638 | |||
NM_022827.3 | NP_073738.2 | 5 | 583 | |||
GenBank mRNA | AK025000.1 | - | 4 | 432 | ||
AK092731.1 | - | 5 | 568 | |||
AK125807.1 | - | 7 | 1313 | |||
AK225189.1 | - | 4 | 432 | |||
AK291805.1 | - | 5 | 526 | |||
AK302850.1 | - | 4 | 482 | |||
AK308616.1 | - | 5 | 708 | |||
AK310416.1 | - | 4 | 427 | |||
BC025255.1 | - | 5 | 471 | 5019010 | ||
BC065526.1 | - | 5 | 525 | 6149633 | ||
BC111029.1 | - | 4 | 440 | 5245538 | ||
BM787432.1 | - | 4 | 498 | |||
CR936769.1 | - | 4 | 453 | |||
DQ238597.1 | - | 4 | 501 | |||
GQ472220.1 | - | 4 | 440 |