Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022830.2 | NP_073741.2 | 5 | 1270 | ||
GenBank mRNA | AB101208.1 | - | 5 | 1184 | ||
AK025920.1 | - | 1 | 233 | |||
AK026000.1 | - | 5 | 1177 | |||
AK292610.1 | - | 5 | 1269 | |||
BC005013.1 | - | 1 | 232 | |||
BC110910.1 | - | 5 | 1159 | 6141937 | ||
BC128263.1 | - | 5 | 1163 | 40118672 | ||
KU178756.1 | - | 1 | 101 | |||
KU178757.1 | - | 1 | 101 |