Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000756.3 | NP_000747.1 | 2 | 1268 | ||
XM_017013090.1 | XP_016868579.1 | 6 | 1606 | |||
XM_017013091.1 | XP_016868580.1 | 6 | 1669 | |||
XM_017013092.1 | XP_016868581.1 | 6 | 1607 | |||
XM_017013093.1 | XP_016868582.1 | 6 | 1746 | |||
GenBank mRNA | AK075431.1 | - | 2 | 1214 | ||
BC011031.1 | - | 2 | 1090 | 4280754 |