Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TTC23 SIRNA Assays ›
Gene Symbol:
TTC23
Gene Name:
tetratricopeptide repeat domain 23
Gene Aliases:
HCC-8
Chromosome Location:
Chr.15: 99136323 - 99251226 on Build GRCh38
UniGene ID:
Hs.513195
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s35098
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TTC23
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001288615.1
NP_001275544.1
9
1526
NM_001288616.1
NP_001275545.1
7
917
NR_109947.1
8
1187
XM_011521927.2
XP_011520229.1
5
667
XM_011521929.1
XP_011520231.1
9
1234
XM_011521930.1
XP_011520232.1
8
1112
XM_011521931.1
XP_011520233.1
7
1255
XM_011521932.1
XP_011520234.1
8
1123
XM_011521933.1
XP_011520235.1
7
1056
XM_011521934.1
XP_011520236.1
9
1526
XM_011521935.1
XP_011520237.1
7
1238
XM_011521936.1
XP_011520238.1
7
917
XM_011521937.1
XP_011520239.1
6
1107
XM_011521939.1
XP_011520241.1
7
983
XM_011521940.2
XP_011520242.1
5
667
XM_011521941.2
XP_011520243.1
5
667
XM_011521943.2
XP_011520245.1
5
667
XM_011521944.2
XP_011520246.1
5
667
XM_011521945.2
XP_011520247.1
5
667
XM_017022509.1
XP_016877998.1
9
1685
XM_017022510.1
XP_016877999.1
8
1348
XM_017022511.1
XP_016878000.1
7
1122
XM_017022512.1
XP_016878001.1
7
1255
XM_017022513.1
XP_016878002.1
8
1123
XM_017022514.1
XP_016878003.1
7
1056
XM_017022515.1
XP_016878004.1
7
1238
XM_017022516.1
XP_016878005.1
7
1122
XM_017022517.1
XP_016878006.1
8
1039
XM_017022518.1
XP_016878007.1
7
983
GenBank mRNA
AF411456.1
-
9
1497
AK022634.1
-
2
102
AK023001.1
-
4
669
AK023230.1
-
9
1225
AK222580.1
-
2
98
AK291690.1
-
7
914
BC015728.1
-
8
1086
4857155
BM755170.1
-
3
241
DA681018.1
-
3
258
Back To Top
More Information
Gene Ontology Categories:
Process(es)
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-cnw2s:80/10.239.163.240:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0