Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321995.1 | NP_001308924.1 | 10 | 938 | ||
NM_001321996.1 | NP_001308925.1 | 10 | 1038 | |||
NM_001321997.1 | NP_001308926.1 | 10 | 859 | |||
NM_031902.4 | NP_114108.1 | 10 | 1117 | |||
NR_136152.1 | 9 | 947 | ||||
NR_136153.1 | 9 | 1026 | ||||
GenBank mRNA | AB049940.1 | - | 10 | 998 | ||
AK058160.1 | - | 9 | 800 | |||
BC000219.2 | - | 6 | 432 | |||
BC014172.2 | - | 10 | 1090 | 4561399 |