Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAMKV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320147.1 | NP_001307076.1 | 8 | 1165 | ||
NM_024046.4 | NP_076951.2 | 8 | 1165 | |||
GenBank mRNA | AK074856.1 | - | 8 | 879 | ||
AK074899.1 | - | 8 | 849 | |||
AK075294.1 | - | 8 | 860 | |||
AK295588.1 | - | 8 | 723 | |||
AK297258.1 | - | 8 | 811 | |||
AK297437.1 | - | 8 | 805 | |||
AK299953.1 | - | 8 | 808 | |||
AK315525.1 | - | 8 | 898 | |||
AL136697.1 | - | 7 | 764 | |||
AL833943.1 | - | 6 | 1223 | |||
BC000497.2 | - | 8 | 834 | 2820734 | ||
BC001921.1 | - | 8 | 834 | 2820734 | ||
BC005828.2 | - | 8 | 834 | 2820734 | ||
BC017363.2 | - | 8 | 836 | 3940018 | ||
BC019256.1 | - | 8 | 829 | 3502894 | ||
BX647074.1 | - | 7 | 1007 | |||
CR533508.1 | - | 6 | 602 |