Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAMKV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320147.1 | NP_001307076.1 | 8 | 1235 | ||
NM_024046.4 | NP_076951.2 | 8 | 1235 | |||
GenBank mRNA | AK074856.1 | - | 8 | 949 | ||
AK074899.1 | - | 8 | 919 | |||
AK075294.1 | - | 8 | 930 | |||
AK295588.1 | - | 8 | 793 | |||
AK297258.1 | - | 8 | 881 | |||
AK297437.1 | - | 8 | 875 | |||
AK299953.1 | - | 8 | 878 | |||
AK315525.1 | - | 8 | 968 | |||
AL136697.1 | - | 7 | 834 | |||
AL833943.1 | - | 6 | 1293 | |||
BC000497.2 | - | 8 | 904 | 2820734 | ||
BC001921.1 | - | 8 | 904 | 2820734 | ||
BC005828.2 | - | 8 | 904 | 2820734 | ||
BC017363.2 | - | 8 | 906 | 3940018 | ||
BC019256.1 | - | 8 | 899 | 3502894 | ||
BX647074.1 | - | 7 | 1077 | |||
CR533508.1 | - | 6 | 672 |