Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRRG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024081.5 | NP_076986.1 | 5 | 689 | ||
XM_006718314.3 | XP_006718377.1 | 5 | 539 | |||
XM_017018275.1 | XP_016873764.1 | 5 | 849 | |||
GenBank mRNA | AF326351.1 | - | 5 | 537 | ||
BC010052.2 | - | 5 | 689 | 3841745 | ||
BC063393.1 | - | 5 | 681 | 5183337 | ||
KU178766.1 | - | 4 | 428 | |||
KU178767.1 | - | 4 | 370 |