Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KDELC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318732.1 | NP_001305661.1 | 8 | 1705 | ||
NM_024089.2 | NP_076994.2 | 7 | 1562 | |||
XM_005254075.2 | XP_005254132.1 | 7 | 1562 | |||
GenBank mRNA | AK074974.1 | - | 8 | 1623 | ||
AK222567.1 | - | 7 | 1484 | |||
AK225103.1 | - | 7 | 1485 | |||
AY358959.1 | - | 7 | 1463 | |||
BC001297.1 | - | 7 | 1515 | 3456869 |