Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRYBA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001886.2 | NP_001877.1 | 4 | 251 | ||
XM_006724140.3 | XP_006724203.1 | 4 | 309 | |||
XM_017028598.1 | XP_016884087.1 | 4 | 293 | |||
GenBank mRNA | BC069404.1 | - | 4 | 251 | 7262159 | |
BC096171.3 | - | 3 | 243 | 40003445 | ||
BC096172.3 | - | 3 | 243 | 40003449 | ||
BC096173.3 | - | 3 | 243 | 40003451 | ||
BC096174.1 | - | 3 | 243 | 40003452 | ||
BF726612.1 | - | 4 | 247 | |||
CR456424.1 | - | 4 | 251 | |||
KM230606.1 | - | 3 | 240 | |||
KM365088.1 | - | 3 | 243 | |||
KM365089.1 | - | 4 | 251 | |||
KM365090.1 | - | 3 | 241 | |||
U59057.1 | - | 4 | 252 |