Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM134A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321109.1 | NP_001308038.1 | 9 | 1658 | ||
NM_001321110.1 | NP_001308039.1 | 9 | 1410 | |||
NM_024293.5 | NP_077269.3 | 9 | 1746 | |||
XM_005246848.2 | XP_005246905.1 | 10 | 1614 | |||
GenBank mRNA | AK074983.1 | - | 9 | 1705 | ||
AK091011.1 | - | 10 | 1237 | |||
AL136758.2 | - | 1 | 293 | |||
AL832843.1 | - | 3 | 750 | |||
AL833630.1 | - | 3 | 561 | |||
BC064950.1 | - | 3 | 802 | |||
BC115412.1 | - | 2 | 251 | |||
BC141851.1 | - | 9 | 1621 |