Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM134A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321109.1 | NP_001308038.1 | 5, 6 | 715 | ||
NM_001321110.1 | NP_001308039.1 | 4, 5 | 572 | |||
NM_024293.5 | NP_077269.3 | 5, 6 | 803 | |||
XM_005246848.2 | XP_005246905.1 | 5, 6 | 776 | |||
GenBank mRNA | AK074983.1 | - | 5, 6 | 762 | ||
AK091011.1 | - | 5, 6 | 399 | |||
BC141851.1 | - | 5, 6 | 678 |