Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ZNF557 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044387.1 | NP_001037852.1 | 7, 8 | 907 | ||
NM_001044388.1 | NP_001037853.1 | 7, 8 | 898 | |||
NM_024341.2 | NP_077317.2 | 7, 8 | 919 | |||
GenBank mRNA | AK095524.1 | - | 7, 8 | 652 | ||
AK297283.1 | - | 7, 8 | 676 | |||
AK315405.1 | - | 7, 8 | 936 | |||
BC003020.1 | - | 7, 8 | 669 | 2822966 | ||
BC031208.1 | - | 7, 8 | 676 | |||
BC058040.1 | - | 7, 8 | 648 |