Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030907.3 | NP_112169.2 | 3 | 747 | ||
XM_011542126.2 | XP_011540428.1 | 3 | 1028 | |||
GenBank mRNA | BC002946.2 | - | 3 | 534 | 3955163 | |
BC008702.1 | - | 3 | 534 | 2966866 |