Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024512.4 | NP_078788.2 | 8 | 1441 | ||
XM_011534110.2 | XP_011532412.1 | 8 | 1230 | |||
GenBank mRNA | AJ308569.2 | - | 8 | 1215 | ||
AK057808.1 | - | 8 | 1283 | |||
AK223440.1 | - | 8 | 1283 | |||
AK314658.1 | - | 8 | 1204 | |||
AK315636.1 | - | 8 | 1224 | |||
BC029118.1 | - | 8 | 1213 | 5183639 |