Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GCC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024523.5 | NP_078799.2 | 1 | 1212 | ||
GenBank mRNA | AF525417.1 | - | 1 | 987 | ||
AK092878.1 | - | 1 | 668 | |||
AK315472.1 | - | 1 | 1063 | |||
BC014100.2 | - | 1 | 1212 | 2988591 | ||
BC078665.1 | - | 1 | 1164 | 5923117 |