Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SPAG16 SIRNA Assays ›
Gene Symbol:
SPAG16
Gene Name:
sperm associated antigen 16
Gene Aliases:
PF20, WDR29
Chromosome Location:
Chr.2: 213284369 - 214410501 on Build GRCh38
UniGene ID:
Hs.743365
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s35856
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SPAG16
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001025436.2
NP_001020607.1
4, 5
503
NM_024532.4
NP_078808.3
4, 5
503
NR_047659.1
6, 7
678
XM_006712746.2
XP_006712809.1
4, 5
454
XM_011511814.1
XP_011510116.1
3, 4
551
XM_011511815.1
XP_011510117.1
4, 5
454
XM_011511816.2
XP_011510118.1
4, 5
454
XM_011511817.1
XP_011510119.1
4, 5
454
XM_011511818.1
XP_011510120.1
4, 5
454
XM_011511819.2
XP_011510121.1
3, 4
1202
XM_011511820.1
XP_011510122.1
4, 5
454
XM_011511824.1
XP_011510126.1
4, 5
454
XM_011511827.1
XP_011510129.1
4, 5
454
XM_011511828.1
XP_011510130.1
4, 5
454
XM_011511829.1
XP_011510131.1
4, 5
454
XM_011511831.1
XP_011510133.1
4, 5
454
XM_011511832.1
XP_011510134.1
4, 5
454
XM_011511835.2
XP_011510137.1
3, 4
546
XM_011511837.2
XP_011510139.1
4, 5
454
XM_017004896.1
XP_016860385.1
5, 6
583
XM_017004897.1
XP_016860386.1
2, 3
3200
XM_017004899.1
XP_016860388.1
4, 5
454
XM_017004900.1
XP_016860389.1
4, 5
445
XM_017004901.1
XP_016860390.1
4, 5
435
GenBank mRNA
AF310672.1
-
4, 5
440
AF426740.1
-
4, 5
440
AF490390.1
-
4, 5
433
AK026377.1
-
4, 5
463
AK095036.1
-
3, 4
551
AK310883.1
-
4, 5
402
AL832962.1
-
4, 5
392
BC009614.1
-
4, 5
464
BC025379.1
-
6, 7
683
4820489
BC067756.1
-
4, 5
490
BC100282.1
-
4, 5
451
CR457359.1
-
4, 5
398
Back To Top
More Information
Panther Classification:
Molecular Function -
microtubule or microtubule-binding cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
sperm axoneme assembly
axoneme assembly
cilium assembly
microtubule sliding
cilium movement involved in cell motility
cell motility in response to calcium ion
protein kinase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-574f4745d9-ftf7j:80/10.239.160.32:80.
git-commit: d20c60ba4e48417b3d41b4fa5195ecf3ea66aacf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.12.0-2024.07.30-1.0