Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF128 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024539.3 | NP_078815.3 | 2 | 701 | ||
NM_194463.1 | NP_919445.1 | 2 | 864 | |||
GenBank mRNA | AF394689.1 | - | 2 | 624 | ||
AK074264.1 | - | 2 | 864 | |||
AK126553.1 | - | 2 | 684 | |||
BC036901.1 | - | 2 | 712 | |||
BC063404.1 | - | 2 | 703 | 5191785 |