Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SAP130 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145928.1 | NP_001139400.1 | 16 | 2407 | ||
NM_024545.3 | NP_078821.2 | 15 | 2302 | |||
XM_005263767.3 | XP_005263824.1 | 16 | 2378 | |||
XM_005263768.3 | XP_005263825.1 | 16 | 2357 | |||
XM_005263769.3 | XP_005263826.1 | 16 | 2278 | |||
XM_005263770.3 | XP_005263827.1 | 16 | 2354 | |||
XM_006712748.3 | XP_006712811.1 | 16 | 2564 | |||
XM_006712749.3 | XP_006712812.1 | 15 | 2459 | |||
XM_017004903.1 | XP_016860392.1 | 16 | 2354 | |||
XM_017004904.1 | XP_016860393.1 | 15 | 2252 | |||
XM_017004905.1 | XP_016860394.1 | 15 | 2249 | |||
GenBank mRNA | AK022823.1 | - | 6 | 924 | ||
AK092234.1 | - | 9 | 1360 | |||
AK315185.1 | - | 16 | 2407 | |||
AL136833.1 | - | 15 | 2300 | |||
AY220791.1 | - | 15 | 2170 | |||
BC017453.1 | - | 8 | 1107 | |||
BC117255.1 | - | 15 | 2268 | 40125806 | ||
BC143898.1 | - | 16 | 2373 | 9052416 |