Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCRN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024583.4 | NP_078859.2 | 2 | 149 | ||
XM_005246853.2 | XP_005246910.1 | 2 | 554 | |||
XM_005246854.4 | XP_005246911.1 | 2 | 1714 | |||
XM_005246855.2 | XP_005246912.1 | 2 | 602 | |||
XM_005246856.2 | XP_005246913.1 | 2 | 558 | |||
XM_005246857.2 | XP_005246914.1 | 2 | 258 | |||
XM_017004909.1 | XP_016860398.1 | 2 | 1146 | |||
XM_017004910.1 | XP_016860399.1 | 2 | 624 | |||
GenBank mRNA | AK057912.1 | - | 2 | 126 | ||
AK296558.1 | - | 2 | 148 | |||
BC119684.1 | - | 2 | 100 | 40116568 | ||
BC119685.1 | - | 2 | 100 | 40116570 | ||
BG492502.1 | - | 2 | 140 | |||
BM725063.1 | - | 1 | 140 | |||
DA760451.1 | - | 2 | 149 |