Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300746.1 | NP_001287675.1 | 2 | 873 | ||
NM_001300747.1 | NP_001287676.1 | 3 | 905 | |||
NM_001300748.1 | NP_001287677.1 | 3 | 902 | |||
NM_024587.3 | NP_078863.2 | 3 | 995 | |||
XM_011542138.2 | XP_011540440.1 | 3 | 1124 | |||
GenBank mRNA | AK026006.1 | - | 3 | 802 | ||
AK225405.1 | - | 3 | 802 | |||
AK295356.1 | - | 2 | 595 | |||
AK296548.1 | - | 2 | 711 | |||
AK310016.1 | - | 3 | 939 | |||
AL832539.1 | - | 2 | 707 | |||
BC064520.1 | - | 3 | 814 | 5582377 | ||
CR457348.1 | - | 3 | 778 |