Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other GTDC1 SIRNA Assays ›
Gene Symbol:
GTDC1
Gene Name:
glycosyltransferase like domain containing 1
Gene Aliases:
Hmat-Xa, mat-Xa
Chromosome Location:
Chr.2: 143945787 - 144333092 on Build GRCh38
UniGene ID:
Hs.44780
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s36156
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: GTDC1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001006636.3
NP_001006637.1
10
1447
NM_001164629.3
NP_001158101.1
10
1432
NM_001284233.1
NP_001271162.1
7
1111
NM_001284234.1
NP_001271163.1
8
1147
NM_001284238.1
NP_001271167.1
12
1646
NM_024659.4
NP_078935.2
8
1159
XM_005263774.2
XP_005263831.1
11
1531
XM_005263782.2
XP_005263839.1
11
1547
XM_005263784.2
XP_005263841.1
9
1406
XM_011511843.2
XP_011510145.1
11
1580
XM_011511855.2
XP_011510157.1
9
1524
XM_011511856.2
XP_011510158.1
9
1306
XM_017004917.1
XP_016860406.1
15
2202
XM_017004918.1
XP_016860407.1
13
1794
XM_017004919.1
XP_016860408.1
11
1680
XM_017004920.1
XP_016860409.1
13
4836
XM_017004921.1
XP_016860410.1
12
1629
XM_017004922.1
XP_016860411.1
12
1806
XM_017004923.1
XP_016860412.1
11
1594
XM_017004924.1
XP_016860413.1
13
1790
XM_017004925.1
XP_016860414.1
10
1452
XM_017004926.1
XP_016860415.1
8
1383
XM_017004927.1
XP_016860416.1
10
1414
XM_017004928.1
XP_016860417.1
12
1647
XM_017004929.1
XP_016860418.1
10
1384
XM_017004930.1
XP_016860419.1
10
2035
XM_017004931.1
XP_016860420.1
10
1448
XM_017004932.1
XP_016860421.1
10
1418
XM_017004933.1
XP_016860422.1
10
1400
XM_017004934.1
XP_016860423.1
11
4607
XM_017004935.1
XP_016860424.1
12
2477
XM_017004936.1
XP_016860425.1
11
1654
XM_017004937.1
XP_016860426.1
8
1390
XM_017004938.1
XP_016860427.1
7
1278
XM_017004939.1
XP_016860428.1
10
1438
XM_017004940.1
XP_016860429.1
7
1239
XM_017004941.1
XP_016860430.1
6
1128
XM_017004942.1
XP_016860431.1
9
1285
XM_017004943.1
XP_016860432.1
9
1951
XM_017004944.1
XP_016860433.1
8
1174
XM_017004945.1
XP_016860434.1
10
1392
XM_017004946.1
XP_016860435.1
8
1129
XM_017004947.1
XP_016860436.1
8
1812
XM_017004948.1
XP_016860437.1
9
4352
XM_017004949.1
XP_016860438.1
7
1028
XM_017004950.1
XP_016860439.1
9
1075
XM_017004951.1
XP_016860440.1
9
1030
XM_017004952.1
XP_016860441.1
9
1474
XM_017004953.1
XP_016860442.1
9
1046
XM_017004954.1
XP_016860443.1
10
1134
XM_017004955.1
XP_016860444.1
12
1482
XM_017004956.1
XP_016860445.1
6
885
XM_017004957.1
XP_016860446.1
8
931
GenBank mRNA
AB201269.1
-
7
1168
AB597184.1
-
7
1168
AK021815.1
-
4
623
AK091684.1
-
7
1115
AK291357.1
-
10
1362
AY281366.1
-
10
1447
BC017741.2
-
9
1199
4390876
BX647436.1
-
8
1129
BX647773.1
-
12
1585
Back To Top
More Information
Panther Classification:
Molecular Function -
glycosyltransferase
transferase
metabolite interconversion enzyme
Gene Ontology Categories:
Process(es)
transferase activity, transferring glycosyl groups
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-n6vvh:80/10.239.163.216:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0