Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other GTDC1 SIRNA Assays ›
Gene Symbol:
GTDC1
Gene Name:
glycosyltransferase like domain containing 1
Gene Aliases:
Hmat-Xa, mat-Xa
Chromosome Location:
Chr.2: 143945787 - 144333092 on Build GRCh38
UniGene ID:
Hs.44780
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s36157
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: GTDC1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001006636.3
NP_001006637.1
12
1608
NM_001164629.3
NP_001158101.1
12
1593
NM_001284233.1
NP_001271162.1
9
1272
NM_001284234.1
NP_001271163.1
10
1308
NM_001284238.1
NP_001271167.1
14
1807
NM_024659.4
NP_078935.2
10
1320
XM_005263774.2
XP_005263831.1
13
1692
XM_005263782.2
XP_005263839.1
13
1708
XM_005263784.2
XP_005263841.1
11
1567
XM_011511843.2
XP_011510145.1
13
1741
XM_011511855.2
XP_011510157.1
11
1685
XM_011511856.2
XP_011510158.1
11
1467
XM_017004917.1
XP_016860406.1
17
2363
XM_017004918.1
XP_016860407.1
15
1955
XM_017004919.1
XP_016860408.1
13
1841
XM_017004920.1
XP_016860409.1
15
4997
XM_017004921.1
XP_016860410.1
14
1790
XM_017004922.1
XP_016860411.1
14
1967
XM_017004923.1
XP_016860412.1
13
1755
XM_017004924.1
XP_016860413.1
15
1951
XM_017004925.1
XP_016860414.1
12
1613
XM_017004926.1
XP_016860415.1
10
1544
XM_017004927.1
XP_016860416.1
12
1575
XM_017004928.1
XP_016860417.1
14
1808
XM_017004929.1
XP_016860418.1
12
1545
XM_017004930.1
XP_016860419.1
12
2196
XM_017004931.1
XP_016860420.1
12
1609
XM_017004932.1
XP_016860421.1
12
1579
XM_017004933.1
XP_016860422.1
12
1561
XM_017004934.1
XP_016860423.1
13
4768
XM_017004935.1
XP_016860424.1
14
2638
XM_017004936.1
XP_016860425.1
13
1815
XM_017004937.1
XP_016860426.1
10
1551
XM_017004938.1
XP_016860427.1
9
1439
XM_017004939.1
XP_016860428.1
12
1599
XM_017004940.1
XP_016860429.1
9
1400
XM_017004941.1
XP_016860430.1
8
1289
XM_017004942.1
XP_016860431.1
11
1446
XM_017004943.1
XP_016860432.1
11
2112
XM_017004944.1
XP_016860433.1
10
1335
XM_017004945.1
XP_016860434.1
12
1553
XM_017004946.1
XP_016860435.1
10
1290
XM_017004947.1
XP_016860436.1
10
1973
XM_017004948.1
XP_016860437.1
11
4513
XM_017004949.1
XP_016860438.1
9
1189
XM_017004950.1
XP_016860439.1
11
1236
XM_017004951.1
XP_016860440.1
11
1191
XM_017004952.1
XP_016860441.1
11
1635
XM_017004953.1
XP_016860442.1
11
1207
XM_017004954.1
XP_016860443.1
12
1295
XM_017004955.1
XP_016860444.1
14
1643
XM_017004956.1
XP_016860445.1
8
1046
XM_017004957.1
XP_016860446.1
10
1092
GenBank mRNA
AB201269.1
-
9
1329
AB597184.1
-
9
1329
AK021815.1
-
6
784
AK091684.1
-
9
1276
AK291357.1
-
12
1523
AY281366.1
-
12
1608
BC017741.2
-
11
1360
4390876
BX647436.1
-
10
1290
BX647773.1
-
14
1746
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001024274, AY281367.This siRNA also targets the following transcripts in Mouse: NM_172662, BC052069, AK162289, AK054272.
Panther Classification:
Molecular Function -
glycosyltransferase
transferase
metabolite interconversion enzyme
Gene Ontology Categories:
Process(es)
transferase activity, transferring glycosyl groups
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-n6vvh:80/10.239.163.216:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0