Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NSUN7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024677.4 | NP_078953.3 | 8 | 1616 | ||
XM_017008613.1 | XP_016864102.1 | 8 | 1639 | |||
XM_017008614.1 | XP_016864103.1 | 8 | 1639 | |||
GenBank mRNA | AK304048.1 | - | 8 | 1340 | ||
BC036568.2 | - | 8 | 1588 | 5272990 | ||
BC042401.1 | - | 8 | 1616 |