Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TXNDC15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024715.3 | NP_078991.3 | 4 | 1442 | ||
GenBank mRNA | AK026278.1 | - | 5 | 845 | ||
AK056512.1 | - | 4 | 1477 | |||
AK092425.1 | - | 2 | 1507 | |||
AK225454.1 | - | 5 | 845 | |||
AK225462.1 | - | 5 | 845 | |||
AK299578.1 | - | 4 | 735 | |||
AK307529.1 | - | 3 | 353 | |||
AY358649.1 | - | 4 | 870 | |||
BC001615.1 | - | 4 | 929 | 3544923 | ||
BC032568.1 | - | 4 | 843 | 5225890 | ||
BX538131.1 | - | 4 | 769 |