Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RERGL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286201.1 | NP_001273130.1 | 4 | 310 | ||
NM_024730.3 | NP_079006.1 | 5 | 467 | |||
GenBank mRNA | AK026308.1 | - | 5 | 393 | ||
BC042888.1 | - | 5 | 454 | 5198817 | ||
BX422875.2 | - | 4 | 258 | |||
DT220095.1 | - | 5 | 467 |