Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CLMN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024734.3 | NP_079010.2 | 9 | 1050 | ||
XM_011537158.1 | XP_011535460.1 | 9 | 1046 | |||
XM_011537159.2 | XP_011535461.1 | 9 | 1046 | |||
XM_017021646.1 | XP_016877135.1 | 9 | 1216 | |||
XM_017021647.1 | XP_016877136.1 | 9 | 1046 | |||
GenBank mRNA | AB047979.1 | - | 9 | 1007 | ||
AK022279.1 | - | 4 | 394 | |||
AK023828.1 | - | 1 | 18 | |||
AK025226.1 | - | 9 | 993 | |||
AK314315.1 | - | 9 | 1048 | |||
AL117526.1 | - | 9 | 902 | |||
BC032699.1 | - | 4 | 369 | |||
BC068482.1 | - | 9 | 999 | 4375469 |